Merck
  • Ezrin-anchored PKA phosphorylates serine 369 and 373 on connexin 43 to enhance gap junction assembly, communication, and cell fusion.
Ezrin-anchored PKA phosphorylates serine 369 and 373 on connexin 43 to enhance gap junction assembly, communication, and cell fusion.
The Biochemical journal (2017-12-21)
Aleksandra R Dukic, Pascale Gerbaud, Jean Guibourdenche, Bernd Thiede, Kjetil Taskén, Guillaume Pidoux
ABSTRACT
A limited number of human cells can fuse to form multinucleated syncytia. In the differentiation of human placenta, mononuclear cytotrophoblasts fuse to form an endocrinologically active, non-proliferative, multinucleated syncytium. This syncytium covers the placenta and manages the exchange of nutrients and gases between maternal and fetal circulation. We recently reported protein kinase A (PKA) to be part of a macromolecular signaling complex with ezrin and gap junction protein connexin 43 (Cx43) that provides cAMP-mediated control of gap junction communication. Here, we examined the associated phosphorylation events. Inhibition of PKA activity resulted in decreased Cx43 phosphorylation, which was associated with reduced trophoblast fusion and differentiation. In vitro studies using peptide arrays, together with mass spectrometry, pointed to serine 369 and 373 of Cx43 as the major PKA phosphorylation sites that increases gap junction assembly at the plasmalemma. A combination of knockdown and reconstitution experiments and gap-fluorescence loss in photobleaching assays with mutant Cx43 containing single or double phosphoserine-mimicking amino acid substitutions in putative PKA phosphorylation sites demonstrated that phosphorylation of S369 and S373 mediated gap junction communication, trophoblast differentiation, and cell fusion.
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit