Merck
  • TRIM21-SERPINB5 aids GMPS repression to protect nasopharyngeal carcinoma cells from radiation-induced apoptosis.
TRIM21-SERPINB5 aids GMPS repression to protect nasopharyngeal carcinoma cells from radiation-induced apoptosis.
Journal of biomedical science (2020-02-02)
Panpan Zhang, Xiaomin Li, Qiuping He, Lulu Zhang, Keqing Song, Xiaojing Yang, Qingmei He, Yaqin Wang, Xiaohong Hong, Jun Ma, Na Liu
ABSTRACT
The main strategy against nasopharyngeal carcinoma (NPC) is radiotherapy. However, radioresistance mediated recurrence is a leading clinical bottleneck in NPC. Revealing the mechanism of NPC radioresistance will help improve the therapeutic effect. In this study, the role of TRIM21 (tripartite motif-containing 21) in NPC receiving ionizing radiation was firstly examined both in vivo and in vitro. Mass spectrometry analysis was performed to identify the downstream targets of TRIM21. NPC cells with TRIM21 or SERPINB5 (serpin family B member 5) overexpression or knockout were used to determine the epistatic relationship among SERPINB5, GMPS (guanine monophosphate synthase) and TRIM21. Flow cytometry, co-immunoprecipitation, western blot and immunofluorescence were employed to strengthen the results. Finally, immunohistochemistry using 4 radiosensitive and 8 radioresistent NPC patient samples was perform to examine the association between SERPINB5 or GMPS expression and patient radio-sensitivity. As an E3 ligase, TRIM21 was highly expressed in NPC. After ionizing radiation, TRIM21 repressed TP53 expression by mediating GMPS ubiquitination and degradation. Overexpression of TRIM21 protected NPC cells from radiation mediated cell apoptosis in vitro and in vivo. Further analysis revealed that TRIM21 mediated GMPS repression was dependent on SERPINB5, and SERPINB5 served as an adaptor which prevented GMPS from entering into the nucleus and introduced TRIM21 for GMPS ubiquitination. Moreover, the in vitro and in vivo results validated the finding that SERPINB5 promoted NPC cell radioresistance, and the radioresistant patients had higher SERPINB5 expression. Overall, our data showed that TRIM21-SERPINB5-mediated GMPS degradation facilitated TP53 repression, which promoted the radioresistance of NPC cells. This novel working model related to TP53 suppression provided new insight into NPC radioresistence clinically.
Sigma-Aldrich
Monoclonal ANTI-FLAG® antibody produced in rabbit, clone SIG1-25, ascites fluid
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)