Merck
  • A simplified aortic ring assay: A useful ex vivo method to assess biochemical and functional parameters of angiogenesis.
A simplified aortic ring assay: A useful ex vivo method to assess biochemical and functional parameters of angiogenesis.
Matrix biology plus (2021-02-06)
Aastha Kapoor, Carolyn G Chen, Renato V Iozzo
ABSTRACT
We present a simplified method for conducting aortic ring assays which yields robust sprouting and high reproducibility targeted towards matrix biologists studying angiogenesis and extracellular matrix signaling. Main adjustments from previously established protocols include embedding aortic rings between two layers of 3D type I collagen matrix and supplementing with vascular endothelial media. We also introduce a concise and effective staining protocol for obtaining high-resolution images of intracellular and extracellular matrix proteins along with a more accurate protocol to quantify angiogenesis. Importantly, we present a novel method to perform biochemical analyses of vessel sprouting without contamination from the aortic ring itself. Overall, our refined method enables detection of low abundance and phosphorylated proteins and provides a straightforward ex vivo angiogenic assay that can be easily reproduced by those in the matrix biology field.
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®
Sigma-Aldrich
Collagen Type I, rat tail, 100 mg, solution, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-p62/SQSTM1 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®