Merck
  • FOXM1 increases hTERT protein stability and indicates poor prognosis in gastric cancer.
FOXM1 increases hTERT protein stability and indicates poor prognosis in gastric cancer.
Neoplasia (New York, N.Y.) (2022-12-19)
Qingyun Tang, Cheng Liu, Shengwei Zhang, Lijiao He, Yunyi Liu, Jing Wang, Xiaoyan Zhao, Xinzhe Li
ABSTRACT
Gastric cancer is one of most lethal diseases across the world. However, the underlying mechanism of gastric cancer carcinogenesis and development is still not fully known. Forkhead box M1 (FOXM1) belongs to the FOX family and has crucial roles in transactivation of multiple oncogenes in several cancer types, including gastric cancer. Recent studies have also shown the non-transcriptional function of FOXM1 via protein-protein interactions. Human telomerase reverse transcriptase (hTERT) is the core subunit of telomerase that facilitates cancer initiation and progression by maintaining cell immortalization, promoting cell proliferation and inhibiting cell apoptosis. However, the relationship between FOXM1 and hTERT in gastric cancer is still unclear. In our study, we found that FOXM1 and hTERT were convergent to the cell cycle-related pathways and they were positively related with advanced gastric cancer stages and poor outcomes. Simultaneous high levels of FOXM1 and hTERT predicted the worst prognosis. FOXM1 could increase hTERT protein rather than mRNA levels in a non-transcriptional manner. Mechanistically, FOXM1 interrupted the interaction between the E3 ligase MKRN1 and hTERT and decreased hTERT protein degradation. Further studies revealed that FOXM1 interacted with hTERT through its DNA-binding domain (DBD) region. Finally, we found that hTERT played important roles in FOXM1-mediated activation of the Wnt/β-catenin pathway to promote gastric cancer cell proliferation. Taken together, we found a novel non-classical function of FOXM1 to increase hTERT protein stability. Targeting the FOXM1-hTERT pathway may be a potential therapeutic strategy in treating gastric cancer.
Product Description
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution