Merck
  • Conformational stability and ligand binding properties of BldR, a member of the MarR family, from Sulfolobus solfataricus.
Conformational stability and ligand binding properties of BldR, a member of the MarR family, from Sulfolobus solfataricus.
Biochimica et biophysica acta (2014-04-08)
Gabriella Fiorentino, Immacolata Del Giudice, Luigi Petraccone, Simonetta Bartolucci, Pompea Del Vecchio
ABSTRACT
The multiple antibiotic resistance regulators (MarR) constitute a family of ligand-responsive transcriptional regulators ubiquitous among the bacterial and archaeal domains. BldR, an archaeal MarR member characterized from the hyperthermophilic crenarchaeon Sulfolobus solfataricus regulates its own expression and that of an alcohol dehydrogenase gene by binding to sequences in their promoters and responding to benzaldehyde as the effector molecule. In this study we assessed the thermodynamic stability of the protein BldR and its binding with benzaldehyde through biophysical measurements. The temperature- and denaturant-induced unfolding experiments, performed by means of circular dichroism (CD) and differential scanning calorimetry (DSC), showed that BldR has an extremely high thermal stability (Td=108.9°C) and a remarkable resistance against GuHCl (Cm=5.3M at 25°C). The unfolding Gibbs energy, ΔdG (H2O), calculated by the linear extrapolation model from GuHCl-induced unfolding equilibrium curve, is 72.2kJmol(-1). ITC binding experiments showed that four benzaldehyde molecules bind to one BldR dimer with a binding constant Kb of 7.5·10(5)M(-1), being the binding entropically driven. ITC, CD and fluorescence results are consistent with a conformational change induced by benzaldehyde binding, further proving that this molecule is a specific effector for BldR modulating its DNA binding activity.
Product Description
Supelco
Benzaldehyde, analytical standard
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Guanidine sulfate salt, 99%
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%
SAFC
Guanidine hydrochloride
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, Molecular Biology, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)