Merck
  • Dietary rapeseed/canola-oil supplementation reduces serum lipids and liver enzymes and alters postprandial inflammatory responses in adipose tissue compared to olive-oil supplementation in obese men.
Dietary rapeseed/canola-oil supplementation reduces serum lipids and liver enzymes and alters postprandial inflammatory responses in adipose tissue compared to olive-oil supplementation in obese men.
Molecular nutrition & food research (2014-11-19)
Michael Kruse, Christian von Loeffelholz, Daniela Hoffmann, Antje Pohlmann, Anne-Cathrin Seltmann, Martin Osterhoff, Silke Hornemann, Olga Pivovarova, Sascha Rohn, Gerhard Jahreis, Andreas F H Pfeiffer
ABSTRACT
Obesity is associated with hyperlipidemia, hepatic steatosis, and low-grade inflammation. Studies have shown that MUFA as well as PUFA have beneficial effects on blood lipids and the inflammatory state. This study investigates the effects of a daily supplementation of either 50 g of rapeseed/canola (RA) or olive (OL) oil over 4 wk on serum lipids, serum liver enzymes, and inflammatory gene expression in subcutaneous (s. c.) adipose tissue in obese men. Consuming RA resulted in increased serum n-3 fatty acids and a reduction in total cholesterol, LDL cholesterol, and serum aspartate aminotransferase compared to OL. In s. c. adipose tissue, gene expression of the pro-inflammatory cytokine IL6 was reduced in RA compared to OL. However, after 4 h after a test meal, containing the appropriate oil, white bread, and 400 mL of liquid diet drink (835 kcal in total), gene expression of IL6, IL1B, and EMR1 (egf-like module containing Mucin-like hormone receptor-like 1) was increased in RA and of monocyte chemoattractant protein-1 (CCL2) in both RA and OL. This demonstrates that consuming RA for 4 wk improves serum lipids, liver enzymes, and basal inflammation in s. c. adipose tissue, but it mediates an acute pro-inflammatory response in adipose tissue upon consuming a meal.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Boron, crystalline, 1 cm, 99.7% trace metals basis
Sigma-Aldrich
Boron, ≥95% (boron), amorphous powder
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Boron, monofilament, 10m, diameter 0.1mm
Boron, monofilament, 20m, diameter 0.2mm
Boron, monofilament, 5m, diameter 0.2mm
Boron, monofilament, 50m, diameter 0.1mm
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Boron, monofilament, 20m, diameter 0.1mm
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Boron, monofilament, 50m, diameter 0.2mm
Boron, monofilament, 10m, diameter 0.2mm
Boron, monofilament, 5m, diameter 0.1mm
Boron, monofilament, 200m, diameter 0.1mm
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes