Merck
  • Direct, simple derivatization of disulfide bonds in proteins with organic mercury in alkaline medium without any chemical pre-reducing agents.
Direct, simple derivatization of disulfide bonds in proteins with organic mercury in alkaline medium without any chemical pre-reducing agents.
Analytica chimica acta (2014-08-26)
Beatrice Campanella, Massimo Onor, Carlo Ferrari, Alessandro D'Ulivo, Emilia Bramanti
ABSTRACT
In this work we have studied the derivatization of protein disulfide bonds with p-Hydroxymercurybenzoate (pHMB) in strong alkaline medium without any preliminary reduction. The reaction has been followed by the determination of the protein-pHMB complex using size exclusion chromatography coupled to a microwave/UV mercury oxidation system for the on-line oxidation of free and protein-complexed pHMB and atomic fluorescence spectrometry (SEC-CVG-AFS) detection. The reaction has been optimized by an experimental design using lysozyme as a model protein and applied to several thiolic proteins. The proposed method reports, for the first time, that it is possible to label 75-100% cysteines of proteins and, thus, to determine thiolic proteins without the need of any reducing step to obtain reduced SH groups before mercury labelling. We obtained a detection limit of 100 nmol L(-1) based on a signal-to-noise ratio of 3 for unbound and complexed pHMB, corresponding to a detection limit of proteins ranged between 3 and 360 nmol L(-1), depending on the number of cysteines in the protein sequence.
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥97%
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Supelco
Benzoic acid, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Guanidine hydrochloride, Molecular Biology, ≥99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
USP
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%
SAFC
Guanidine hydrochloride