Merck
  • Effect of telmisartan on VEGF-induced and VEGF-independent angiogenic responsiveness of coronary endothelial cells in normal and streptozotocin (STZ)-induced diabetic rats.
Effect of telmisartan on VEGF-induced and VEGF-independent angiogenic responsiveness of coronary endothelial cells in normal and streptozotocin (STZ)-induced diabetic rats.
Clinical and experimental hypertension (New York, N.Y. : 1993) (2014-02-05)
Kiranj K Chaudagar, Anita A Mehta
ABSTRACT
Telmisartan possesses endothelial protective effects due to angiotensin II type 1 receptor antagonist, peroxisome proliferator-activated receptor γ (PPARγ) agonist and antioxidant action. Therefore, our objective was to study effect of telmisartan on angiogenic responsiveness of coronary endothelial cells (cECs) of normal and diabetic rats. Male Wistar rats were divided into six groups, normal rats, diabetic rats 30 d. (30 days after administration of STZ), diabetic rats 60 ds. (60 days after administration of STZ), telmisartan-treated normal rats (2 mg/kg, p.o., for 15 days before isolation of hearts), telmisartan-treated diabetic rats 30 ds, and telmisartan-treated diabetic rats 60 ds. Each group was further divided into two subgroups, sham rat hearts and ischemia-reperfused rat hearts. After isolation of cEC from each subgroup, angiogenic responsiveness and nitric oxide releasing properties were studied using chorioallantoic membrane (CAM) assay and Griess method, respectively. cEC of normal rats showed significant increase in angiogenic responsiveness in presence of vascular endothelial growth factor (VEGF) but not in absence of it. This activity was attenuated by pretreatment of cEC with l-NAME, wortmannin and chelerythrine. Diabetes and ischemia reperfusion injury suppressed angiogenic responsiveness of cEC. Telmisartan treatment showed significant increase in VEGF-induced angiogenic responsiveness and nitric oxide releasing properties of cECs of all subgroups as compared to their respective non-treated subgroups. These effects of telmisartan were significantly inhibited by pretreatment of cECs with L-NAME and wortmannin but not with chelerythrine. Our data suggest that telmisartan improves VEGF-induced coronary angiogenic activity in normal and diabetic rats via stimulation of PI3K/eNOS/NO pathway.
Product Description
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications