Merck
  • Simvastatin reduces leucocyte- and platelet-endothelial cell interaction in murine antigen-induced arthritis in vivo.
Simvastatin reduces leucocyte- and platelet-endothelial cell interaction in murine antigen-induced arthritis in vivo.
Scandinavian journal of rheumatology (2014-05-16)
O Gottschalk, M L Dao Trong, P Metz, J Wallmichrath, S Piltz, K W Jauch, V Jansson, M Schmitt-Sody
ABSTRACT
The use of statins in the prevention and treatment of cardiovascular diseases is well established. Their use as anti-inflammatory and immunomodulatory agents in the treatment of rheumatoid arthritis (RA) has also been investigated, with several clinical and experimental studies indicating an anti-inflammatory effect of statins for RA, but other studies showing no effect or even the opposite. The current study was designed to examine the effect of simvastatin in an in vivo murine model of arthritis using intravital microscopy. We assigned four groups (n = 7, female C57Bl6 mice), two with and two without antigen-induced arthritis (AiA), from which one of the non-AiA groups and one of the AiA groups were treated with simvastatin 40 mg/kg i.p. daily for 14 consecutive days after induction of arthritis. Platelet- and leucocyte-endothelial cell interaction was assessed by measurement of rolling and adherent fluorescence-labelled platelets and leucocytes, functional capillary density (FCD) was evaluated, and knee joint diameter was determined as a clinical parameter. In arthritic mice treated with simvastatin, a significant reduction in platelet- and leucocyte-endothelial cell interaction was observed in comparison to arthritic mice treated with vehicle. In addition, a significant reduction in FCD was seen in arthritic mice treated with simvastatin, along with a reduction in knee joint swelling of the AiA mice. Treatment of AiA mice with simvastatin showed significant reductions in platelet- and leucocyte-endothelial cell interactions, in FCD, and in the swelling of the knee joint. These results support the hypothesis of the anti-inflammatory effects of statins in the treatment of RA.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Supelco
Simvastatin, Pharmaceutical Secondary Standard; Certified Reference Material
Alprostadil, European Pharmacopoeia (EP) Reference Standard
USP
Simvastatin, United States Pharmacopeia (USP) Reference Standard
Supelco
Simvastatin, analytical standard
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Simvastatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Prostaglandin E1, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E1, ≥98% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E1, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)