Merck
  • Selective microemulsion liquid chromatography analysis of dopamine receptor antagonist LE300 and its N-methyl metabolite in mouse sera by using a monolithic silica column.
Selective microemulsion liquid chromatography analysis of dopamine receptor antagonist LE300 and its N-methyl metabolite in mouse sera by using a monolithic silica column.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-03-31)
Abdulrhman A Al-Majed, Mohamed M Hefnawy, Mostafa S Mohammed, Sabry M Attia, Jochen Lehmann
ABSTRACT
A highly selective, sensitive, and rapid microemulsion liquid chromatography (MELC) method was developed and validated for the simultaneous determination of a novel type of dopamine receptor antagonist LE300 and its N-methyl metabolite in mouse sera. LE300, its N-methyl metabolite, and pindolol (an internal standard) were detected using excitation and emission wavelengths of 275 and 340 nm, respectively. HPLC analysis by using a monolithic column was performed by directly injecting the sample after appropriate dilution with the microemulsion mobile phase. The chromatographic behaviour of these compounds was studied to demonstrate their chromatographic efficiency, retention, and peak symmetry. The MELC method was validated for its specificity, linearity, accuracy, precision, robustness and stability. An experimental design was used during validation to evaluate method robustness. The calibration curves in serum showed excellent linearity (r=0.997) over concentrations ranging from 10 to 400 ngmL(-1) for LE300 and 15 to 500 ngmL(-1) for its N-methyl metabolite. The mean relative standard deviation (RSD) of the results of inter- and intra-day precision and accuracy of LE300 and its N-methyl metabolite were ≤5%. The overall recoveries of LE300 and its N-methyl metabolite from mouse sera were in the range 97.9-101.5% with %RSD ranging from 0.98% to 3.63%, which were in line with ICH guidelines. The assay was successfully applied in a pharmacokinetic study.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Pindolol, European Pharmacopoeia (EP) Reference Standard
Supelco
Diisopropyl ether, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder
Sigma-Aldrich
Diisopropyl ether, anhydrous, 99%, contains either BHT or hydroquinone as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Diisopropyl ether, ReagentPlus®, 99%, contains either BHT or hydroquinone as stabilizer
Sigma-Aldrich
Diisopropyl ether, contains either BHT or hydroquinone as stabilizer, ACS reagent, ≥99.0%
Sigma-Aldrich
Diisopropyl ether, puriss. p.a., ≥98.5% (GC)
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%