Merck
  • Human metabolic responses to chronic environmental polycyclic aromatic hydrocarbon exposure by a metabolomic approach.
Human metabolic responses to chronic environmental polycyclic aromatic hydrocarbon exposure by a metabolomic approach.
Journal of proteome research (2015-05-21)
Zhonghua Wang, Yajie Zheng, Baoxin Zhao, Yanping Zhang, Zhe Liu, Jing Xu, Yanhua Chen, Zhao Yang, Fenfen Wang, Huiqing Wang, Jiuming He, Ruiping Zhang, Zeper Abliz
ABSTRACT
The toxicities of polycyclic aromatic hydrocarbons (PAHs) have been extensively explored due to their carcinogenic and mutagenic potency; however, little is known about the metabolic responses to chronic environmental PAH exposure among the general population. In the present study, 566 healthy volunteers were dichotomized into exposed and control groups to investigate PAH-induced perturbations in the metabolic profiles. Nine urine PAH metabolites were measured by a sensitive LC-MS/MS method to comprehensively evaluate the PAH exposure level of each individual, and the metabolic profiles were characterized via a LC-MS-based metabolomic approach. PAH exposure was correlated to its metabolic outcomes by linear and logistic regression analyses. Metabolites related to amino acid, purine, lipid, and glucuronic acid metabolism were significantly changed in the exposed group. 1-Hydroxyphenanthrene and dodecadienylcarnitine have potential as sensitive and reliable biomarkers for PAH exposure and its metabolic outcomes, respectively, in the general population. These findings generally support the hypothesis that environmental PAH exposure causes oxidative stress-related effects in humans. The current study provides new insight into the early molecular events induced by PAH exposure in the actual environment.
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Azelaic acid, 98%
Sigma-Aldrich
DL-Phenylalanine, ReagentPlus®, 99%
Sigma-Aldrich
Azelaic acid, technical grade, 80%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2-Naphthol, 99%
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Sigma-Aldrich
3-Methyl-L-histidine
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Naphthol, 98%
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
2-Hydroxyfluorene, 98%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
Kynurenic acid, ≥98%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
Progesterone, meets USP testing specifications