Merck
  • Protective Effects of Melatonin against Cyclophosphamide-induced Oxidative Lung Toxicity in Mice.
Protective Effects of Melatonin against Cyclophosphamide-induced Oxidative Lung Toxicity in Mice.
Drug research (2014-03-29)
M Shokrzadeh, A Chabra, F Naghshvar, A Ahmadi, M Jafarinejhad, Y Hasani-Nourian
ABSTRACT
This study was undertaken to evaluate the protective effects of melatonin against cyclophosphamide (CP)-induced oxidative lung toxicity in mice. Mice were pre-treated with various doses of melatonin for 7 consecutive days and were then injected with CP (200 mg/kg b. w.) 1 h after last melatonin injection. After 24 h, the mice were euthanized and their lungs were immediately harvested. Several biomarkers associated with oxidative stress in lung homogenates, such as thiobarbituric acid reactive substances (TBARs) and reduced glutathione (GSH) levels and the activity of superoxide dismutase (SOD) and catalase (CAT) were measured spectrophotometrically. A single dose of CP markedly altered the levels of these oxidative stress biomarkers in lung homogenates. However, increased lipid peroxidation, measured as TBARs, was significantly inhibited in the lung tissues of the melatonin-pretreated mice compared to the CP alone-injected group. In addition, pretreatment with melatonin also significantly restored GSH levels and SOD and CAT activities. Melatonin also effectively protected animals from CP-induced histological abnormalities in lung tissue. In conclusion, the increase in oxidative stress markers and concomitant adaptations by the antioxidant defense system indicates that oxidative stress plays an important role in CP-induced damage to the lung. Moreover, melatonin is a potent natural antioxidant that helps prevent CP-induced oxidative toxicity in mouse lung tissues. Thus, because melatonin is regarded to be a safe pineal secretory product, it may be used concomitantly as a supplement to reduce lung damage in patients undergoing chemotherapy.
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Iron(III) chloride, ≥99.9% trace metals basis
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%