Merck
  • Phase I clinical trial of 99mTc-etarfolatide, an imaging agent for folate receptor in healthy Japanese adults.
Phase I clinical trial of 99mTc-etarfolatide, an imaging agent for folate receptor in healthy Japanese adults.
Annals of nuclear medicine (2015-08-05)
Yoshinori Yamada, Hiroshi Nakatani, Hisashi Yanaihara, Masahiro Omote
ABSTRACT
Technetium etarfolatide ((99m)Tc-EF) is a radioactive diagnostic imaging agent that was developed to assess the expression of folate receptors in tumors. Administering folic acid prior to the administration of (99m)Tc-EF has been shown to improve SPECT images. Here, we conducted a phase I clinical trial to assess the safety, pharmacokinetics, and radiation dosimetry of (99m)Tc-EF injection following pre-administration of folic acid in healthy Japanese male adults. Six healthy Japanese male adults were enrolled in the study. Folic acid was intravenously administered, followed 1-3 min later by an intravenous injection of (99m)Tc-EF (740 MBq ± 20 %). Assessments of subjective symptoms and objective findings, electrocardiograms, physical examination, and laboratory tests were performed before and up to 7 days after the injection to assess the safety of (99m)Tc-EF. Blood and urine collections and whole-body planar imaging were conducted at various time points up to 24 h after the injection to assess the pharmacokinetics of (99m)Tc-EF. The internal radiation dosimetry was calculated based on the pharmacokinetics results using the MIRD method. Five adverse events were observed in three subjects (50 %) after administration of the folic acid and (99m)Tc-EF, while these events were mild and non-serious. Of those five events, three were considered to be related to the administered agents. The radioactivity in blood rapidly decreased and showed a biphasic profile. The activity of (99m)Tc-EF at 5 min post injection was largest in the bone marrow, followed by the liver and kidneys, and had decreased within 24 h in all organs/tissues without appreciable retention. The pharmacokinetics results suggested that (99m)Tc-EF was mainly eliminated by kidney. The results also suggested that when administered at 925 MBq of (99m)Tc-EF, which is the maximum dose generally used for clinical trials in other countries, the corresponding effective dose of (99m)Tc-EF is equal to or less than those determined for the current radioactive diagnostic imaging agents. The results of this study assessing the safety and radiation dosimetry of (99m)Tc-EF with folic acid pre-administration suggested that folic acid and (99m)Tc-EF should be appropriate for further studies. No pharmacokinetics concerns were noted.
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Folic acid, ≥97%