Merck
  • Electrodeposition of reduced graphene oxide on a Pt electrode and its use as amperometric sensor in microchip electrophoresis.
Electrodeposition of reduced graphene oxide on a Pt electrode and its use as amperometric sensor in microchip electrophoresis.
Electrophoresis (2015-04-18)
Bruno Gabriel Lucca, Fábio de Lima, Wendell K T Coltro, Valdir Souza Ferreira
ABSTRACT
This report describes the development and application of a novel graphene-modified electrode to be used as amperometric sensor in microchip electrophoresis (ME) devices. The modified electrode was achieved based on electroreduction of graphene oxide on an integrated Pt working electrode of a commercial ME device. The surface modification was characterized by SEM and cyclic voltammetry techniques. The results indicated that graphene sheets were successfully deposited exhibiting higher surface conductivity and greater electrode sensitivity. The performance of the modified electrode for the amperometric detection on ME devices has been demonstrated by the separation and detection of an anionic mixture containing iodide and ascorbate. The graphene-modified electrode provided significantly higher sensitivity (896.7 vs. 210.9 pA/μM for iodide and 217.8 vs. 127.8 pA/μM for ascorbate), better separation efficiencies (3400 vs. 700 plates/m for iodide and 10 000 vs. 2400 plates/m for ascorbate), enhanced peak resolutions (1.6 vs. 1.0), and LODs (1.5 vs. 5.3 μM for iodide and 3.1 vs. 7.3 μM for ascorbate) in comparison with the unmodified Pt electrode. The proposed amperometric sensor was successfully applied for the analysis of ascorbic acid (through its anionic form) in a commercial medicine sample, and the results achieved were in agreement with the value provided by the supplier. Based on the data here presented, the modified graphene electrode shows great promise for ME applications.
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥97%
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis