Merck
  • X-FISH: Analysis of cellular RNA expression patterns using flow cytometry.
X-FISH: Analysis of cellular RNA expression patterns using flow cytometry.
Journal of immunological methods (2015-05-23)
Aja M Rieger, Jeffrey J Havixbeck, Daniel R Barreda
ABSTRACT
Fluorescent in situ hybridization (FISH) is a powerful technique for the detection of RNA or DNA within cells and tissues, which provides a unique link between molecular and cell biology. This technique is broadly applicable across a range of biological systems. While FISH has been previously adapted to flow-based platforms, their use remains limited because of procedural challenges and costs associated with commercial kits. Herein we present a protocol that modifies existing techniques to sensitively and specifically detect and examine RNA expression patterns in primary cells and cell lines using flow cytometry (expression-FISH; X-FISH). As relevant examples, we show how this technique can be used to monitor changes in mRNA expression following activation, how it can be combined with antibody staining to study RNA and protein in the same sample, and how it can help distinguish among subsets in a mixed cell population. X-FISH can integrate multiple probes and can be performed in conjunction with other assays, allowing for informative multiparametric analyses and increased statistical robustness. For non-classical comparative animal models this procedure provides a time saving alternative to de novo production of antibody-based markers. Finally, X-FISH provides an economical solution that is applicable to conventional as well as multi-spectral imaging flow cytometry platforms.
Product Description
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder