Merck
  • Posttranslational modifications of intact proteins detected by NMR spectroscopy: application to glycosylation.
Posttranslational modifications of intact proteins detected by NMR spectroscopy: application to glycosylation.
Angewandte Chemie (International ed. in English) (2015-05-01)
Mario Schubert, Michal J Walczak, Markus Aebi, Gerhard Wider
ABSTRACT
Posttranslational modifications (PTMs) are an integral part of the majority of proteins. The characterization of structure and function of PTMs can be very challenging especially for glycans. Existing methods to analyze PTMs require complicated sample preparations and suffer from missing certain modifications, the inability to identify linkage types and thus chemical structure. We present a direct, robust, and simple NMR spectroscopy method for the detection and identification of PTMs in proteins. No isotope labeling is required, nor does the molecular weight of the studied protein limit the application. The method can directly detect modifications on intact proteins without sophisticated sample preparation. This approach is well suited for diagnostics of proteins derived from native organisms and for the quality control of biotechnologically produced therapeutic proteins.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D