Merck
  • Pilot-scale chitin extraction from shrimp shell waste by deproteination and decalcification with bacterial enrichment cultures.
Pilot-scale chitin extraction from shrimp shell waste by deproteination and decalcification with bacterial enrichment cultures.
Applied microbiology and biotechnology (2015-08-01)
Mini Bajaj, Andrea Freiberg, Josef Winter, Youmei Xu, Claudia Gallert
ABSTRACT
Extraction of chitin from mechanically pre-purified shrimp shells can be achieved by successive NaOH/HCl treatment, protease/HCl treatment or by environmentally friendly fermentation with proteolytic/lactic acid bacteria (LAB). For the last mentioned alternative, scale-up of shrimp shell chitin purification was investigated in 0.25 L (F1), 10 L (F2), and 300 L (F3) fermenters using an anaerobic, chitinase-deficient, proteolytic enrichment culture from ground meat for deproteination and a mixed culture of LAB from bio-yoghurt for decalcification. Protein removal in F1, F2, and F3 proceeded in parallel within 40 h at an efficiency of 89-91 %. Between 85 and 90 % of the calcit was removed from the shells by LAB in another 40 h in F1, F2, and F3. After deproteination of shrimp shells in F3, spent fermentation liquor was re-used for a next batch of 30-kg shrimp shells in F4 (300 L) which eliminated 85.5 % protein. The purity of the resulting chitin was comparable in F1, F2, F3, and F4. Viscosities of chitosan, obtained after chitin deacetylation and of chitin, prepared biologically or chemically in the laboratory, were much higher than those of commercially available chitin and chitosan.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether