Merck
  • Cyclohexane-1,2-dicarboxylic acid diisononyl ester and metabolite effects on rat epididymal stromal vascular fraction differentiation of adipose tissue.
Cyclohexane-1,2-dicarboxylic acid diisononyl ester and metabolite effects on rat epididymal stromal vascular fraction differentiation of adipose tissue.
Environmental research (2015-04-13)
Enrico Campioli, Tam B Duong, François Deschamps, Vassilios Papadopoulos
ABSTRACT
Plastics are generally mixed with additives like plasticizers to enhance their flexibility, pliability, and elasticity proprieties. Plasticizers are easily released into the environment and are absorbed mainly through ingestion, dermal contact, and inhalation. One of the main classes of plasticizers, phthalates, has been associated with endocrine and reproductive diseases. In 2002, 1,2-cyclohexane dicarboxylic acid diisononyl ester (DINCH) was introduced in the market for use in plastic materials and articles intended to come into contact with food, and it received final approval from the European Food Safety Authority in 2006. At present, there is limited knowledge about the safety and potential metabolic and endocrine-disrupting properties of DINCH and its metabolites. The purpose of this study was to evaluate the biological effects of DINCH and its active metabolites, cyclohexane-1,2-dicarboxylic acid (CHDA) and cyclohexane-1,2-dicarboxylic acid mono isononyl ester (MINCH), on rat primary stromal vascular fraction (SVF) of adipose tissue. DINCH and its metabolite, CHDA, were not able to directly affect SVF differentiation. However, exposure of SVF to 50 μM and 100 μM concentrations of MINCH affected the expression of Cebpa and Fabp4, thus inducing SVF preadipocytes to accumulate lipids and fully differentiate into mature adipocytes. The effect of MINCH was blocked by the specific peroxisome proliferator-activated receptor (PPAR)-α antagonist, GW6471. Taken together, these results suggest that MINCH is a potent PPAR-α agonist and a metabolic disruptor, capable of inducing SVF preadipocyte differentiation, that may interfere with the endocrine system in mammals.
Sigma-Aldrich
T0070907, ≥98% (HPLC)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
SAFC
BIS-TRIS
Sigma-Aldrich
GW6471, ≥98% (HPLC)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
SAFC
BIS-TRIS
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder