Merck
  • ARF Confers a Context-Dependent Response to Chemotherapy in Muscle-Invasive Bladder Cancer.
ARF Confers a Context-Dependent Response to Chemotherapy in Muscle-Invasive Bladder Cancer.
Cancer research (2017-01-14)
Tomasz B Owczarek, Takashi Kobayashi, Ricardo Ramirez, Lijie Rong, Anna M Puzio-Kuter, Gopa Iyer, Min Yuen Teo, Francisco Sánchez-Vega, Jingqiang Wang, Nikolaus Schultz, Tian Zheng, David B Solit, Hikmat A Al-Ahmadie, Cory Abate-Shen
ABSTRACT
Muscle-invasive bladder cancer (MIBC) generally responds poorly to treatment and tends to exhibit significant mortality. Here we show that expression of the tumor suppressor p14
Sigma-Aldrich
MISSION® esiRNA, targeting human ITGB3BP
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)