Our company is pleased to partner with ChiroSolve, Inc. to offer, through an exclusive distribution agreement, a series of ready-to-use kits for chiral resolution of racemates. These kits allow scientists to quickly screen resolving agents and solvents against a racemate to find the optimum combination, as well as to optimize reaction conditions in order to separate a racemic mixture into its constituent enantiomers. They can be used for racemic acids, bases, alcohols, amino acids, aldehydes, and ketones.
There are three Acid Series kits and three Base Series kits, available individually or as the complete acid or base set. Each individual kit is in a ready-to-use, 96-vial, high-throughput format that allows the entire analysis to be performed inside the kit without removing any vials. The kit is made of polypropylene material that can withstand extreme temperatures (–20 to 120 °C), allowing the entire kit to be placed in an oven or ice bath without damage.
Each vial has calibrated quantities of a unique resolving agent and solvent combination. Resolving agents are chosen with manufacturing use in mind; they are relatively inexpensive and recoverable in high yield after the resolution is complete. Using all three kits in an acid or base series, 288 resolving agent–solvent combinations can be explored for a single racemate. The high-throughput format allows scientists to identify within one day the optimum resolution process which might otherwise take over 2 months.
Acid |
Base |
Acid Series 1, 2, and 3 include a distinct group of chirally pure acids and are used to resolve racemic bases and amino acids (some pre-processing is required for amino acids). Examples of resolving agents in the acid kits are: |
Base Series 1, 2, and 3 include a distinct group of chirally pure bases or amines and are used to resolve racemic acids, alcohols, aldehydes, and ketones (some pre-processing is required for the latter three). Examples of resolving agents in the base kits are: |
|
|
ChiroSolv Kits have been used by several pharmaceutical, fine chemical, and contract R&D companies to reduce their product development time and increase speed to market. Two examples of the application of this technology for determining the ideal solvent/resolving agent combination for the resolution of key compounds are shown.
In the first case, a key advanced intermediate in the synthesis of (S)-(–)-cetirizine, a non-sedative histamine H-1 receptor antagonist used for the treatment of allergies, was screened against the kits in the Acid Series. The combination of l-(+)-tartaric acid in methanol was determined to provide the maximum optical purity after its chiral resolution.
Case Study 1
In the second example, (–)-ephedrine in isobutanol was determined to be the ideal combination for resolving indoline-2-carboxylic acid to yield the (S)-(–)-enantiomer, which is an intermediate for ACE inhibitors such as trandolapril and perindopril. For this compound, the racemic mixture was screened against the three ChiroSolv Kits in the Base Series.
Case Study 2
ChiroSolv Kits use the method of diastereomeric crystallization, a process that chemically separates enantiomers in a racemic mixture by complexation with an enantiopure acid or base, resulting in a mixture of diastereomeric salts. These salts have different chemical and physical properties, which allow their separation. This wellestablished technology competes favorably with newer techniques such as asymmetric synthesis, biocatalysis, enzyme resolution, kinetic resolution, or chiral chromatography.
One major advantage of diastereomeric crystallization is that the procedure scales up easily for manufacturing purposes. Today around 65% of chiral products are made using this technique. Examples: