Seleccione un Tamaño
| Talla/SKU | Disponibilidad | Precio |
|---|---|---|
1 unit | Póngase en contacto con nuestro Servicio de Atención al Cliente para disponibilidad | 745,00 € |
Acerca de este artículo
745,00 €
material
aluminum
description
removable
manufacturer/tradename
Milliflex®
application(s)
bioburden testing
food and beverages
pharmaceutical
water monitoring
compatibility
for use with Milliflex® Quantum Reader System
General description
The drawer can be removed easily and replaced without any specific tools.
The Milliflex® Quantum Reader is used to detect viable microbes in filtered samples through a rapid fluorescence-based technology from food and pharma samples.
Application
Legal Information
1 of 1
Este artículo | |||
|---|---|---|---|
| description For Milliflex® Quantum Reader | description pkg of 48 tests, suitable for bioburden testing | description Part of the Milliflex® Quantum system, Fluorescence-based technology, For the rapid detection of microbial contamination | description with validation protocol A4 format, For the rapid detection of microbial contamination |
| compatibility for use with Milliflex® Quantum Reader System | compatibility - | compatibility - | compatibility - |
| material aluminum | material - | material polyester housing, aluminum housing (sheet 1050), PVC foot, 304L stainless steel chamber (optical), 304L stainless steel handle, transparent polycarbonate protector(s) (LED), glass filter (OG550; optical) | material - |
| application(s) bioburden testing | application(s) bioburden testing | application(s) pharmaceutical | application(s) pharmaceutical |
| manufacturer/tradename Milliflex® | manufacturer/tradename Milliflex® Quantum | manufacturer/tradename Milliflex® | manufacturer/tradename Milliflex® |
| description removable | description - | description - | description - |
Elija entre una de las versiones más recientes:
¿Ya tiene este producto?