Merck
  • A Mad2-Mediated Translational Regulatory Mechanism Promoting S-Phase Cyclin Synthesis Controls Origin Firing and Survival to Replication Stress.
A Mad2-Mediated Translational Regulatory Mechanism Promoting S-Phase Cyclin Synthesis Controls Origin Firing and Survival to Replication Stress.
Molecular cell (2018-05-19)
Sophie Gay, Daniele Piccini, Christopher Bruhn, Sara Ricciardi, Paolo Soffientini, Walter Carotenuto, Stefano Biffo, Marco Foiani
RESUMEN
Cell survival to replication stress depends on the activation of the Mec1ATR-Rad53 checkpoint response that protects the integrity of stalled forks and controls the origin firing program. Here we found that Mad2, a member of the spindle assembly checkpoint (SAC), contributes to efficient origin firing and to cell survival in response to replication stress. We show that Rad53 and Mad2 promote S-phase cyclin expression through different mechanisms: while Rad53 influences Clb5,6 degradation, Mad2 promotes their protein synthesis. We found that Mad2 co-sediments with polysomes and modulates the association of the translation inhibitor Caf204E-BP with the translation machinery and the initiation factor eIF4E. This Mad2-dependent translational regulatory process does not depend on other SAC proteins. Altogether our observations indicate that Mad2 has an additional function outside of mitosis to control DNA synthesis and collaborates with the Mec1-Rad53 regulatory axis to allow cell survival in response to replication stress.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cicloheximida, from microbial, ≥94% (TLC)
Sigma-Aldrich
Anticuerpo anti-fosfo-histona H2A.X (Ser139), clon JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Microesferas de vidrio lavadas con ácido, 425-600 μm (30-40 U.S. sieve)
Sigma-Aldrich
Methyl methanesulfonate, 99%
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
(R)-MG132
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Camptothecin, Camptotheca acuminata, A cell-permeable DNA topoisomerase I inhibitor.