Merck
RNA polymerase II: subunit structure and function.
Trends in biochemical sciences (1990-09-01)
N A Woychik, R A Young
RESUMEN
RNA polymerase II is the core of the complex apparatus that is responsible for the regulated synthesis of mRNA. A comprehensive knowledge of RNA polymerase II is essential to our understanding of the molecular mechanisms through which a variety of transcription factors regulate eukaryotic gene expression. The recent cloning of genes for all ten subunits of yeast RNA polymerase II has revealed intriguing similarities and differences between the eukaryotic RNA polymerase and its simpler prokaryotic counterpart. Epitope tagging and other experiments made possible by the cloning of these genes have provided a clearer picture of RNA polymerase II subunit composition, stoichiometry and function, and set the stage for further investigating the dialogue between RNA polymerase II and transcription factors.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
RNA Polymerase II, RPB10 subunit, GST tagged human, recombinant, expressed in E. coli, ≥85% (SDS-PAGE)