Merck
  • Selective Prostacyclin Receptor Agonist Selexipag, in Contrast to Prostacyclin Analogs, Does Not Evoke Paradoxical Vasoconstriction of the Rat Femoral Artery.
Selective Prostacyclin Receptor Agonist Selexipag, in Contrast to Prostacyclin Analogs, Does Not Evoke Paradoxical Vasoconstriction of the Rat Femoral Artery.
The Journal of pharmacology and experimental therapeutics (2018-03-29)
Keith Morrison, Franck Haag, Roland Ernst, Marc Iglarz, Martine Clozel
RESUMEN
Selexipag [2-{4-[(5,6-diphenylpyrazin-2-yl)(isopropyl)amino]butoxy}-N-(methylsulfonyl)acetamide] is a selective nonprostanoid prostacyclin (PGI2) receptor (IP receptor) agonist that is approved for the treatment of pulmonary arterial hypertension (PAH). In contrast to selexipag, PGI2 analogs used in the clinic are nonselective agonists at prostanoid receptors and can also activate contractile prostaglandin E receptor 3 (EP3) receptors. Leg pain is a common side effect in patients receiving treatment with PGI2 analogs and peripheral vasoconstriction can be responsible for side effects related to muscular ischemia. This study tested the hypothesis that PGI2 analogs could cause paradoxical vasoconstriction of the femoral artery via EP3 receptor activation but that only vasorelaxation would be observed in response to selexipag and its active metabolite ACT-333679 [{4-[(5,6-diphenylpyrazin-2-yl)(isopropyl)amino]butoxy}acetic acid]. Selexipag and ACT-333679 relaxed rings of the isolated rat femoral artery contracted with either prostaglandin F2 α (PGF2 α ) or the α1 adrenoceptor (α1AR) agonist phenylephrine. ACT-333679 also inhibited contraction of the femoral artery to sympathetic nerve stimulation. In contrast, PGI2 analogs (iloprost, beraprost, and treprostinil) caused additional contraction of arterial rings precontracted with phenylephrine, which was reverted to relaxation by antagonism of EP3 receptors. Treprostinil augmented contraction of the femoral artery to sympathetic nerve stimulation in an EP3 receptor-dependent manner. Mechanistically, concomitant EP3 and α1AR receptor activation synergistically constricted femoral arteries. It is concluded that selexipag and ACT-333679 are vasorelaxants of the rat femoral artery and, unlike PGI2 analogs, do not cause paradoxical vasoconstriction via activation of EP3 receptors. EP3 receptor-mediated vasoconstriction may contribute to the well documented peripheral muscle pain reported in patients with PAH receiving PGI2 analogs. Leg pain may be less in patients treated with selexipag.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Sulprostone, ≥95% (HPLC), oil