Merck
Lymphatic exosomes promote dendritic cell migration along guidance cues.
The Journal of cell biology (2018-04-14)
Markus Brown, Louise A Johnson, Dario A Leone, Peter Majek, Kari Vaahtomeri, Daniel Senfter, Nora Bukosza, Helga Schachner, Gabriele Asfour, Brigitte Langer, Robert Hauschild, Katja Parapatics, Young-Kwon Hong, Keiryn L Bennett, Renate Kain, Michael Detmar, Michael Sixt, David G Jackson, Dontscho Kerjaschki
RESUMEN
Lymphatic endothelial cells (LECs) release extracellular chemokines to guide the migration of dendritic cells. In this study, we report that LECs also release basolateral exosome-rich endothelial vesicles (EEVs) that are secreted in greater numbers in the presence of inflammatory cytokines and accumulate in the perivascular stroma of small lymphatic vessels in human chronic inflammatory diseases. Proteomic analyses of EEV fractions identified >1,700 cargo proteins and revealed a dominant motility-promoting protein signature. In vitro and ex vivo EEV fractions augmented cellular protrusion formation in a CX3CL1/fractalkine-dependent fashion and enhanced the directional migratory response of human dendritic cells along guidance cues. We conclude that perilymphatic LEC exosomes enhance exploratory behavior and thus promote directional migration of CX3CR1-expressing cells in complex tissue environments.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
5-Carboxytetramethylrhodamine
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)