Merck
IQGAP1 binds the Axl receptor kinase and inhibits its signaling.
The Biochemical journal (2018-09-07)
Laëtitia Gorisse, Zhigang Li, Andrew C Hedman, David B Sacks
RESUMEN
Axl is a tyrosine kinase receptor that is important for hematopoiesis, the innate immune response, platelet aggregation, engulfment of apoptotic cells and cell survival. Binding of growth arrest-specific protein 6 (Gas6) activates Axl signaling, but the mechanism of inactivation of the Axl receptor is poorly understood. In the present study, we show that IQGAP1 modulates Axl signaling. IQGAP1 is a scaffold protein that integrates cell signaling pathways by binding several growth factor receptors and intracellular signaling molecules. Our in vitro analysis revealed a direct interaction between the IQ domain of IQGAP1 and Axl. Analysis by both immunoprecipitation and proximity ligation assays demonstrated an association between Axl and IQGAP1 in cells and this interaction was decreased by Gas6. Unexpectedly, reducing IQGAP1 levels in cells significantly enhanced the ability of Gas6 to stimulate both Axl phosphorylation and activation of Akt. Moreover, IQGAP1 regulates the interaction of Axl with the epidermal growth factor receptor. Our data identify IQGAP1 as a previously undescribed suppressor of Axl and provide insight into regulation of Axl function.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Reactivos rojos para detección in situ Duolink®
Sigma-Aldrich
Sonda para PLA® in situ Duolink® anti-conejo PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® Kit iniciador rojo in situ ratón o conejo
Sigma-Aldrich
Sonda para PLA® in situ Duolink® anti-ratón MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Sonda para PLA® in situ Duolink® anti-conejo MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat