Merck
  • Targeting intestinal epithelial cell-programmed necrosis alleviates tissue injury after intestinal ischemia/reperfusion in rats.
Targeting intestinal epithelial cell-programmed necrosis alleviates tissue injury after intestinal ischemia/reperfusion in rats.
The Journal of surgical research (2018-04-02)
Xiang Li, Yihong Ling, Zhongming Cao, Jiantong Shen, Shaoqian Chen, Weifeng Liu, Baolong Yuan, Shihong Wen
RESUMEN
Intestinal dysfunction, especially acute pathologies linked to intestinal ischemia/reperfusion (I/R) injury, is profoundly affected by inflammation and improper execution of cell death. Few studies have examined the efficacy of combined strategies in regulated intestinal epithelial necrosis after intestinal I/R. Here, we evaluated the functional interaction between poly (adenosine diphosphate-ribose) polymerase 1 (PARP-1)-induced parthanatos and receptor-interacting protein 1/3 (RIP1/3) kinase-induced necroptosis in the pathophysiological course of acute ischemic intestinal injury. Anesthetized adult male Sprague-Dawley rats were subjected to superior mesenteric artery occlusion consisting of 1.5 h of ischemia and 6 h of reperfusion. The PARP-1-specific inhibitor PJ34 (10 mg/kg) and the RIP1-specific inhibitor Necrostatin-1 (1 mg/kg) were intraperitoneally administered 30 min before the induction of ischemia. Intestinal I/R was found to result in PARP-1 activation and RIP1/3-mediated necrosome formation. PJ34 or Necrostatin-1 treatment significantly improved the mucosal injury, while the combined inhibition of PARP-1 and RIP1/3 conferred optimal protection of the intestine. Meanwhile, results from terminal deoxynucleotidyl transferase-mediated deoxyuridine triphosphate-biotin nick end labeling assay showed a decrease in intestinal epithelial cell death. Interestingly, we further showed that PARP-1 might act as a downstream signaling molecule of RIP1 in the process of I/R-induced intestinal injury and that the RIP1/PARP-1-dependent cell death signaling pathway functioned independently of caspase 3 inhibition. The results of our study provide a molecular basis for combination therapy that targets both pathways of regulated necrosis (parthanatos and necroptosis), to treat acute intestinal I/R-induced intestinal epithelial barrier disruption.
MATERIALES
Número de producto
Marca
Descripción del producto
Roche
Kit de detección de muerte celular in situ, POD, sufficient for ≤50 tests
Sigma-Aldrich
Anti-RIPK1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution