Merck
Comparison of protein surface hydrophobicity measured at various pH values using three different fluorescent probes
Journal of Agricultural and Food Chemistry (2000)
Alizadeh-Pasdar N and Li-Chan ECY