Merck
Laser-mediated, site-specific inactivation of RNA transcripts.
Proceedings of the National Academy of Sciences of the United States of America (1999-05-26)
D Grate, C Wilson
RESUMEN
The biological function of specific gene products often is determined experimentally by blocking their expression in an organism and observing the resulting phenotype. Chromophore-assisted laser inactivation using malachite green (MG)-tagged antibodies makes it possible to inactivate target proteins in a highly restricted manner, probing their temporally and spatially resolved functions. In this report, we describe the isolation and in vitro characterization of a MG-binding RNA motif that may enable the same high-resolution analysis of gene function specifically at the RNA level (RNA-chromophore-assisted laser inactivation). A well-defined asymmetric internal bulge within an RNA duplex allows high affinity and high specificity binding by MG. Laser irradiation in the presence of low concentrations of MG induces destruction of the MG-binding RNA but not of coincubated control RNA. Laser-induced hydrolysis of the MG-binding RNA is restricted predominantly to a single nucleotide within the bulge. By appropriately incorporating this motif into a target gene, transcripts generated by the gene may be effectively tagged for laser-mediated destruction.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Adipic acid dihydrazide–Agarose, saline suspension