Merck
DNA methylation-dependent chromatin fiber compaction in vivo and in vitro: requirement for linker histone.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2001-12-01)
M A Karymov, M Tomschik, S H Leuba, P Caiafa, J Zlatanova
RESUMEN
Dynamic alterations in chromatin structure mediated by postsynthetic histone modifications and DNA methylation constitute a major regulatory mechanism in DNA functioning. DNA methylation has been implicated in transcriptional silencing, in part by inducing chromatin condensation. To understand the methylation-dependent chromatin structure, we performed atomic force microscope (AFM) studies of fibers isolated from cultured cells containing normal or elevated levels of m5C. Chromatin fibers were reconstituted on control or methylated DNA templates in the presence or absence of linker histone. Visual inspection of AFM images, combined with quantitative analysis of fiber structural parameters, suggested that DNA methylation induced fiber compaction only in the presence of linker histones. This conclusion was further substantiated by biochemical results.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Sephacryl®, 500-HR, MW range 40-20,000 kDa (dextrans)