Merck
SNW1 is a prognostic biomarker in prostate cancer.
Diagnostic pathology (2019-05-03)
Doris Höflmayer, Carla Willich, Claudia Hube-Magg, Ronald Simon, Dagmar Lang, Emily Neubauer, Frank Jacobsen, Andrea Hinsch, Andreas M Luebke, Marie Christina Tsourlakis, Hartwig Huland, Markus Graefen, Alexander Haese, Hans Heinzer, Sarah Minner, Franziska Büscheck, Guido Sauter, Thorsten Schlomm, Stefan Steurer, Till S Clauditz, Eike Burandt, Waldemar Wilczak, Christian Bernreuther
RESUMEN
SNW1 is a nuclear receptor co-activator involved in splicing and transcription control, including androgen receptor signaling. Overexpression of SNW1 has been linked to adverse prognosis in different cancer types, but studies on the role of SNW1 in prostate cancer are lacking. Using immunohistochemistry, we analyzed SNW1 expression in 10,310 prostate cancers in a tissue microarray (TMA) with attached clinical and molecular data. The comparison with normal prostate tissue revealed an up regulation of SNW1 in a subset of cancer samples. SNW1 staining was considered weak in 31.5%, moderate in 37.7% and strong in 14% of cancers. Strong SNW1 expression was markedly more frequent in prostate cancers harboring the TMPRSS2:ERG fusion (24%) than in ERG negative cancers (7%, p < 0.0001). Significant associations with Gleason grade, stage, nodal status and early biochemical recurrence were observed in the ERG negative and positive subset. Multivariable modeling revealed that the prognostic value of SNW1 up regulation was independent from the established preoperative histopathological and clinical parameters. These results demonstrate that SNW1 overexpression is an independent prognostic marker in prostate cancer with potential clinical utility.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-SNW1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution