Merck
  • Blockade of TNF receptor superfamily 1 (TNFR1)-dependent and TNFR1-independent cell death is crucial for normal epidermal differentiation.
Blockade of TNF receptor superfamily 1 (TNFR1)-dependent and TNFR1-independent cell death is crucial for normal epidermal differentiation.
The Journal of allergy and clinical immunology (2018-03-30)
Xuehua Piao, Ryosuke Miura, Sanae Miyake, Sachiko Komazawa-Sakon, Masato Koike, Ryodai Shindo, Junji Takeda, Akito Hasegawa, Riichiro Abe, Chiharu Nishiyama, Tetsuo Mikami, Hideo Yagita, Yasuo Uchiyama, Hiroyasu Nakano
RESUMEN
A delicate balance between cell death and keratinocyte proliferation is crucial for normal skin development. Previous studies have reported that cellular FLICE (FADD-like ICE)-inhibitory protein plays a crucial role in prevention of keratinocytes from TNF-α-dependent apoptosis and blocking of dermatitis. However, a role for cellular FLICE-inhibitory protein in TNF-α-independent cell death remains unclear. We investigated contribution of TNF-α-dependent and TNF-α-independent signals to the development of dermatitis in epidermis-specific Cflar-deficient (CflarE-KO) mice. We examined the histology and expression of epidermal differentiation markers and inflammatory cytokines in the skin of CflarE-KO;Tnfrsf1a+/- and CflarE-KO;Tnfrsf1a-/- mice. Mice were treated with neutralizing antibodies against Fas ligand and TNF-related apoptosis-inducing ligand to block TNF-α-independent cell death of CflarE-KO;Tnfrsf1a-/- mice. CflarE-KO;Tnfrsf1a-/- mice were born but experienced severe dermatitis and succumbed soon after birth. CflarE-KO;Tnfrsf1a+/- mice exhibited embryonic lethality caused by massive keratinocyte apoptosis. Although keratinocytes from CflarE-KO;Tnfrsf1a-/- mice still died of apoptosis, neutralizing antibodies against Fas ligand and TNF-related apoptosis-inducing ligand substantially prolonged survival of CflarE-KO;Tnfrsf1a-/- mice. Expression of inflammatory cytokines, such as Il6 and Il17a was increased; conversely, expression of epidermal differentiation markers was severely downregulated in the skin of CflarE-KO;Tnfrsf1a-/- mice. Treatment of primary keratinocytes with IL-6 and, to a lesser extent, IL-17A suppressed expression of epidermal differentiation markers. TNF receptor superfamily 1 (TNFR1)-dependent or TNFR1-independent apoptosis of keratinocytes promotes inflammatory cytokine production, which subsequently blocks epidermal differentiation. Thus blockade of both TNFR1-dependent and TNFR1-independent cell death might be an alternative strategy to treat skin diseases when treatment with anti-TNF-α antibody alone is not sufficient.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-α-tubulina monoclonal antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Necrostatin-1, ≥98% (HPLC)