Merck
  • Baculovirus production of fully-active phosphoinositide 3-kinase alpha as a p85alpha-p110alpha fusion for X-ray crystallographic analysis with ATP competitive enzyme inhibitors.
Baculovirus production of fully-active phosphoinositide 3-kinase alpha as a p85alpha-p110alpha fusion for X-ray crystallographic analysis with ATP competitive enzyme inhibitors.
Protein expression and purification (2010-05-12)
Robert H Sinnamon, Patrick McDevitt, Beth L Pietrak, Vaughan R Leydon, Yu Xue, Ruth Lehr, Hongwei Qi, Matthew Burns, Patricia Elkins, Paris Ward, Giorgia Vincentini, Donald Fisher, Maggie Grimes, Martin Brandt, Kurt R Auger, Thau Ho, Kyung Johanson, Christopher S Jones, Benjamin Schwartz, Thomas D Sweitzer, Robert B Kirkpatrick
RESUMEN
Phosphoinositide 3-kinases have been targeted for therapeutic research because they are key components of a cell signaling cascade controlling proliferation, growth, and survival. Direct activation of the PI3Kalpha pathway contributes to the development and progression of solid tumors in breast, endometrial, colon, ovarian, and gastric cancers. In the context of a drug discovery effort, the availability of a robust crystallographic system is a means to understand the subtle differences between ATP competitive inhibitor interactions with the active site and their selectivity against other PI3Kinase enzymes. To generate a suitable recombinant design for this purpose, a p85alpha-p110alpha fusion system was developed which enabled the expression and purification of a stoichiometrically homogeneous, constitutively active enzyme for structure determination with potent ATP competitive inhibitors (Raha et al., in preparation) [56]. This approach has yielded preparations with activity and inhibition characteristics comparable to those of the full-length PI3Kalpha from which X-ray diffracting crystals were grown with inhibitors bound in the active site.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
PI 3-Kinase HTRF® Assay; 1920 wells, The PI 3-Kinase HTRF Assay is a high-performance assay kit that provides a universal method for testing all Class I PI3-Kinases in a homogeneous format. This pack size has a screening capacity of 5 plates.