Merck
MicroRNA-1251-5p Promotes Carcinogenesis and Autophagy via Targeting the Tumor Suppressor TBCC in Ovarian Cancer Cells.
Molecular therapy : the journal of the American Society of Gene Therapy (2019-07-07)
Yang Shao, Xiaomin Liu, Jiao Meng, Xiaofei Zhang, Zhongliang Ma, Gong Yang
RESUMEN
Accounting for more than 70% of ovarian cancer cases, epithelial ovarian malignancy has a low 5-year survival rate. MicroRNAs may be targeted in the clinical treatment of the disease. In this study, we first found that miR-1251-5p was significantly upregulated in human ovarian cancer cell lines and tissues with the cancer progression and stages. Overexpression or inhibition of miR-1251-5p promoted or impeded cell proliferation and cell cycle progression. Subsequently, TBCC, one of the tubulin-binding cofactors (TBCs), was identified as a target of miR-1251-5p to be negatively associated with cell cycle and autophagy. Exogenous overexpression of TBCC inhibited the expressions of CDK4 and LC3BII, but it promoted the expressions of α/β-tubulin and p62 to suppress cell growth and autophagy, particularly under the starving condition; whereas the introduction of miR-1251-5p in TBCC-overexpressing cells rescued the suppressive effects of TBCC on cell cycle and autophagy through the inverse regulation of the above proteins. Finally, miR-1251-5p was proven to enhance xenograft tumor growth through the downregulation of TBCC but upregulation of Ki67 and LC3B in xenograft tumor tissues. Collectively, these results suggest that miR-1251-5p functions as an oncogene to suppress TBCC and α/β-tubulin expression. Thus, the miR-1251-5p/TBCC/α/β-tubulin axis may be targeted for ovarian cancer treatment.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Supelco
Cyanuric acid, analytical standard