Merck
G protein-coupled receptor kinase 5 modifies cancer cell resistance to paclitaxel.
Molecular and cellular biochemistry (2019-08-01)
Joann Lagman, Paula Sayegh, Christina S Lee, Sarah M Sulon, Alec Z Jacinto, Vanessa Sok, Natalie Peng, Deniz Alp, Jeffrey L Benovic, Christopher H So
RESUMEN
G protein-coupled receptor kinases (GRKs) phosphorylate the activated forms of G protein-coupled receptors (GPCRs), leading to receptor desensitization and internalization. In addition, GRKs can modify the activity of many non-GPCR-signaling pathways as well, controlling other cellular functions beyond that directly associated with a GPCR. In this report, we show that cervical cancer HeLa cells and breast cancer MDA MB 231 cells with reduced GRK5 expression display increased sensitivity to the apoptotic effects of paclitaxel (Taxol). This effect in cancer cells with low GRK5 levels could be because of blunted histone deacetylase 6 (HDAC6) activity that leads to an increase in α-tubulin acetylation levels, which augments paclitaxel sensitivity. We demonstrate that GRK5 and HDAC6 form a signaling complex in cells and in vitro. GRK5 phosphorylates HDAC6 at Ser-21 to promote its deacetylase activity. Therefore, the GRK5-HDAC6 interaction may contribute to paclitaxel resistance in cancer cells.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human GRK5