Merck
The roles of multiple importins for nuclear import of murine aristaless-related homeobox protein.
The Journal of biological chemistry (2009-06-06)
Wenbo Lin, Wenduo Ye, Lanlan Cai, Xinyi Meng, Guifen Ke, Caoxin Huang, Zi Peng, Yinhua Yu, Jeffrey A Golden, Alan M Tartakoff, Tao Tao
RESUMEN
Nuclear import of proteins with nuclear localization signals (NLSs) is mediated by shuttling carriers, the importins. Some cargoes display more than a single NLS, and among these are homeodomain proteins such as Arx, which is critical for development of multiple tissues. Arx has two functional NLSs. The present studies show that several pathways can import Arx via its NLS2, which is within its DNA binding homeodomain. Using an in vitro nuclear import assay, we show that import of Arx via NLS2 can be mediated by importin beta1, importin 9, or importin 13, with binding being strongest to importin beta1. All binding is sensitive to RanGTP. Experiments based on precise domain deletions indicate that NLS2 binds impbeta1, imp9, and imp13 and includes both an importin binding subdomain and a regulatory subdomain with arginine residues being important for function. Moreover, Arx can be co-precipitated with these importins when NLS2 is present. Although nuclear import of Arx can be mediated by these three importin betas, importin beta1 seems to play the major role judging from in vivo small interfering RNA ablations and the in vitro import assay. This is the first evidence to show the role of importin beta1 in nuclear import of paired-type homeodomain proteins. We propose a novel and possibly quite general mechanism for nuclear import of paired-type homeodomain proteins which is critical for development.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cóctel de inhibidores de proteasas, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Digitonina, ~50% (TLC)