Merck
A Compendium of Genetic Modifiers of Mitochondrial Dysfunction Reveals Intra-organelle Buffering.
Cell (2019-11-16)
Tsz-Leung To, Alejandro M Cuadros, Hardik Shah, Wendy H W Hung, Yang Li, Sharon H Kim, Daniel H F Rubin, Ryan H Boe, Sneha Rath, John K Eaton, Federica Piccioni, Amy Goodale, Zohra Kalani, John G Doench, David E Root, Stuart L Schreiber, Scott B Vafai, Vamsi K Mootha
RESUMEN
Mitochondrial dysfunction is associated with a spectrum of human conditions, ranging from rare, inborn errors of metabolism to the aging process. To identify pathways that modify mitochondrial dysfunction, we performed genome-wide CRISPR screens in the presence of small-molecule mitochondrial inhibitors. We report a compendium of chemical-genetic interactions involving 191 distinct genetic modifiers, including 38 that are synthetic sick/lethal and 63 that are suppressors. Genes involved in glycolysis (PFKP), pentose phosphate pathway (G6PD), and defense against lipid peroxidation (GPX4) scored high as synthetic sick/lethal. A surprisingly large fraction of suppressors are pathway intrinsic and encode mitochondrial proteins. A striking example of such "intra-organelle" buffering is the alleviation of a chemical defect in complex V by simultaneous inhibition of complex I, which benefits cells by rebalancing redox cofactors, increasing reductive carboxylation, and promoting glycolysis. Perhaps paradoxically, certain forms of mitochondrial dysfunction may best be buffered with "second site" inhibitors to the organelle.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
ANTI-FLAG® M2 monoclonal antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Antimycin A from Streptomyces sp.
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
L-Glutatión reducido, ≥98.0%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, ≥97% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Anti-β-actina monoclonal antibody produced in mouse, clone AC-15, ascites fluid
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
D-(+)-Glucosa solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Oligomicina A, ≥99% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
2-Desoxi-D-glucosa, ≥98% (GC), crystalline
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Rotenone, ≥95%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
(±)-α-Tocoferol, synthetic, ≥96% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
βDinucleótido de β-nicotinamida y adenina sodium salt
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
L-Glutationa oxidada, ≥98% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
MitoTEMPO, ≥98% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ferrostatin-1, ≥95% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Chloramphenicol-Water Soluble, powder, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
BAM15, ≥98% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
D-(+)-Galactosa, Pharmaceutical Secondary Standard; Certified Reference Material
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
L-Glutamine-13C5, 98 atom % 13C, 95% (CP)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
ML 210, ≥98% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Caspase Inhibitor VI, Z-VAD-FMK, CAS 161401-82-7, is an irreversible pan caspase inhibitor. Does not require pretreatment with esterase for in vitro studies.
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Anti-ATP5B antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Anti-G6PD antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Anti-PFKP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
En este momento no podemos mostrarle ni los precios ni la disponibilidad