Merck
Cross-communication between Gi and Gs in a G-protein-coupled receptor heterotetramer guided by a receptor C-terminal domain.
BMC biology (2018-03-01)
Gemma Navarro, Arnau Cordomí, Marc Brugarolas, Estefanía Moreno, David Aguinaga, Laura Pérez-Benito, Sergi Ferre, Antoni Cortés, Vicent Casadó, Josefa Mallol, Enric I Canela, Carme Lluís, Leonardo Pardo, Peter J McCormick, Rafael Franco
RESUMEN
G-protein-coupled receptor (GPCR) heteromeric complexes have distinct properties from homomeric GPCRs, giving rise to new receptor functionalities. Adenosine receptors (A1R or A2AR) can form A1R-A2AR heteromers (A1-A2AHet), and their activation leads to canonical G-protein-dependent (adenylate cyclase mediated) and -independent (β-arrestin mediated) signaling. Adenosine has different affinities for A1R and A2AR, allowing the heteromeric receptor to detect its concentration by integrating the downstream Gi- and Gs-dependent signals. cAMP accumulation and β-arrestin recruitment assays have shown that, within the complex, activation of A2AR impedes signaling via A1R. We examined the mechanism by which A1-A2AHet integrates Gi- and Gs-dependent signals. A1R blockade by A2AR in the A1-A2AHet is not observed in the absence of A2AR activation by agonists, in the absence of the C-terminal domain of A2AR, or in the presence of synthetic peptides that disrupt the heteromer interface of A1-A2AHet, indicating that signaling mediated by A1R and A2AR is controlled by both Gi and Gs proteins. We identified a new mechanism of signal transduction that implies a cross-communication between Gi and Gs proteins guided by the C-terminal tail of the A2AR. This mechanism provides the molecular basis for the operation of the A1-A2AHet as an adenosine concentration-sensing device that modulates the signals originating at both A1R and A2AR.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-Adenosine Receptor A2a Antibody, clone 7F6-G5-A2, clone 7F6-G5-A2, Upstate®, from mouse