Merck
Copper(II)-induced self-oligomerization of alpha-synuclein.
The Biochemical journal (1999-06-09)
S R Paik, H J Shin, J H Lee, C S Chang, J Kim
RESUMEN
alpha-Synuclein is a component of the abnormal protein depositions in senile plaques and Lewy bodies of Alzheimer's disease (AD) and Parkinson's disease respectively. The protein was suggested to provide a possible nucleation centre for plaque formation in AD via selective interaction with amyloid beta/A4 protein (Abeta). We have shown previously that alpha-synuclein has experienced self-oligomerization when Abeta25-35 was present in an orientation-specific manner in the sequence. Here we examine this biochemically specific self-oligomerization with the use of various metals. Strikingly, copper(II) was the most effective metal ion affecting alpha-synuclein to form self-oligomers in the presence of coupling reagents such as dicyclohexylcarbodi-imide or N-(ethoxycarbonyl)-2-ethoxy-1,2-dihydroquinoline. The size distribution of the oligomers indicated that monomeric alpha-synuclein was oligomerized sequentially. The copper-induced oligomerization was shown to be suppressed as the acidic C-terminus of alpha-synuclein was truncated by treatment with endoproteinase Asp-N. In contrast, the Abeta25-35-induced oligomerizations of the intact and truncated forms of alpha-synuclein were not affected. This clearly indicated that the copper-induced oligomerization was dependent on the acidic C-terminal region and that its underlying biochemical mechanism was distinct from that of the Abeta25-35-induced oligomerization. Although the physiological or pathological relevance of the oligomerization remains currently elusive, the common outcome of alpha-synuclein on treatment with copper or Abeta25-35 might be useful in understanding neurodegenerative disorders in molecular terms. In addition, abnormal copper homoeostasis could be considered as one of the risk factors for the development of disorders such as AD or Parkinson's disease.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Endoproteinase Asp-N from Pseudomonas fragi mutant strain, suitable for protein sequencing, lyophilized powder