Merck
Anti-tumoral activity of L and D isomers of aspartic acid beta-hydroxamate on L5178Y leukemia.
International journal of cancer (1991-09-30)
N Thomasset, F Hamedi-Sangsari, R Tournaire, C Navarro, S Malley, L Goetsch, J Grange, J Vila
RESUMEN
D and L isomers of aspartic acid beta-hydroxamate (respectively DAH and LAH) were compared for their in vitro and in vivo activity against the murine leukemia L5178Y and their tolerance in vivo in DBA/2 mice. DAH and LAH displayed comparable cytotoxic activity against L5178Y leukemia in vitro. Death of leukemia cells was observed at concentrations above 1.2 mM for both DAH and LAH. High concentrations of L-asparagine partially reversed the growth-inhibitory effects of DAH and LAH on L5178Y cells for concentrations of DAH and LAH lower than 0.6 mM. Intraperitoneal administration of DAH and LAH to mice showed that the LD10, LD50 and LD90 of DAH was 3- to 4-fold greater for DAH than for LAH. DAH was able to eradicate L5178Y tumors in mice without inducing toxic deaths, whereas LAH at comparable doses killed all the animals treated.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
L-Aspartic acid β-hydroxamate, serine racemase inhibitor