Merck
Modeling of Compressive Strength for Unidirectional Fiber Reinforced Composites with Nanoparticle Modified Epoxy Matrix.
Materials (Basel, Switzerland) (2019-11-30)
Wei Chen, Yiping Liu, Zhenyu Jiang, Liqun Tang, Zejia Liu, Licheng Zhou
RESUMEN
Incorporation of nanoparticles into polymer matrix was found to considerably improve the compressive performance of unidirectional fiber reinforced composites. In our experimental study, an increase by 62.7% in the longitudinal compressive strength of unidirectional carbon fiber reinforced composites is attained by dispersing 8.7 vol.% SiO2 nanoparticles into epoxy matrix. A compressive strength model is established to quantitatively describe the reinforcing effects of nanoparticles, which combines a modified microbuckling model for unidirectional fiber reinforced composites and a constitutive model for nanocomposite matrices under compression. In the two models, the coupling of damage and plasticity is considered to contribute to the nonlinear response of nanocomposite matrix. The proposed strength model demonstrates excellent prediction capability in experimental verification. A small relative deviation below 8.2% is achieved between the predicted compressive strength of unidirectional fiber reinforced composites and the measured values, which is at the same level of random error in experiments.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Bis(4-hydroxyphenyl)methane, 98%