Merck
  • High doses of ultraviolet-C irradiation increases vasoactive intestinal contractor/endothelin-2 expression in keratinocytes of the newborn mouse epidermis.
High doses of ultraviolet-C irradiation increases vasoactive intestinal contractor/endothelin-2 expression in keratinocytes of the newborn mouse epidermis.
Peptides (2007-04-24)
Javier Adur, Satoshi Takizawa, Tsuyoshi Uchide, Victor Casco, Kaname Saida
RESUMEN
We examined the expression profiles of vasoactive intestinal contractor/endothelin-2 (VIC/ET-2) at both gene and peptide level in skin irradiated with different ultraviolet wavelengths. We found that VIC/ET-2 gene expression is sensitive only to ultraviolet-C (UVC) irradiation and has an immediate response. These results provide direct evidence that high doses of UVC irradiation induce an increase in gene expression and protein production of VIC/ET-2 and endothelin (ET) receptors in a dose-dependent manner in epidermal keratinocytes. We suggest that VIC/ET-2 can play an essential role in the maintenance, protection and hyperpigmentation of the epidermis exposed to UVC irradiation from artificial or natural sources.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Vasoactive Intestinal Constrictor / β Endothelin mouse