Merck
Impaired lipid biosynthesis hinders anti-tumor efficacy of intratumoral iNKT cells.
Nature communications (2020-01-25)
Sicheng Fu, Kaixin He, Chenxi Tian, Hua Sun, Chenwen Zhu, Shiyu Bai, Jiwei Liu, Qielan Wu, Di Xie, Ting Yue, Zhuxia Shen, Qingqing Dai, Xiaojun Yu, Shu Zhu, Gang Liu, Rongbin Zhou, Shengzhong Duan, Zhigang Tian, Tao Xu, Hua Wang, Li Bai
RESUMEN
Dysfunction of invariant natural killer T (iNKT) cells in tumor microenvironment hinders their anti-tumor efficacy, and the underlying mechanisms remain unclear. Here we report that iNKT cells increase lipid biosynthesis after activation, and that is promoted by PPARγ and PLZF synergically through enhancing transcription of Srebf1. Among those lipids, cholesterol is required for the optimal IFN-γ production from iNKT cells. Lactic acid in tumor microenvironment reduces expression of PPARγ in intratumoral iNKT cells and consequently diminishes their cholesterol synthesis and IFN-γ production. Importantly, PPARγ agonist pioglitazone, a thiazolidinedione drug for type 2 diabetes, successfully restores IFN-γ production in tumor-infiltrating iNKT cells from both human patients and mouse models. Combination of pioglitazone and alpha-galactosylceramide treatments significantly enhances iNKT cell-mediated anti-tumor immune responses and prolongs survival of tumor-bearing mice. Our studies provide a strategy to augment the anti-tumor efficacy of iNKT cell-based immunotherapies via promoting their lipid biosynthesis.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-phospho-LCK (pTyr394) antibody produced in rabbit, affinity isolated antibody