Merck
  • Potent human glutaminyl cyclase inhibitors as potential anti-Alzheimer's agents: Structure-activity relationship study of Arg-mimetic region.
Potent human glutaminyl cyclase inhibitors as potential anti-Alzheimer's agents: Structure-activity relationship study of Arg-mimetic region.
Bioorganic & medicinal chemistry (2018-02-06)
Van T H Ngo, Van-Hai Hoang, Phuong-Thao Tran, Jihyae Ann, Minghua Cui, Gyungseo Park, Sun Choi, Jiyoun Lee, Hee Kim, Hee-Jin Ha, Kwanghyun Choi, Young-Ho Kim, Jeewoo Lee
RESUMEN
Pyroglutamate-modified amyloid β peptides (pGlu-Aβ) are highly neurotoxic and promote the formation of amyloid plaques. The pGlu-Aβ peptides are generated by glutaminyl cyclase (QC), and recent clinical studies indicate that QC represents an alternative therapeutic target to treat Alzheimer's disease (AD). We have previously developed a series of QC inhibitors with an extended pharmacophoric scaffold, termed the Arg-mimetic D-region. In the present study, we focused on the structure activity relationship (SAR) of analogues with modifications in the D-region and evaluated their biological activity. Most compounds in this series exhibited potent activity in vitro, and our SAR analysis and the molecular docking studies identified compound 202 as a potential candidate because it forms an additional hydrophobic interaction in the hQC active site. Overall, our study provides valuable insights into the Arg-mimetic pharmacophore that will guide the design of novel QC inhibitors as potential treatments for AD.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
tert-Butyl 4-(3-(hydroxymethyl)phenyl)piperidine-1-carboxylate, ≥95%