Merck
  • Induction of CEMIP in Chondrocytes by Inflammatory Cytokines: Underlying Mechanisms and Potential Involvement in Osteoarthritis.
Induction of CEMIP in Chondrocytes by Inflammatory Cytokines: Underlying Mechanisms and Potential Involvement in Osteoarthritis.
International journal of molecular sciences (2020-05-06)
Takashi Ohtsuki, Omer F Hatipoglu, Keiichi Asano, Junko Inagaki, Keiichiro Nishida, Satoshi Hirohata
RESUMEN
In patients with osteoarthritis (OA), there is a decrease in both the concentration and molecular size of hyaluronan (HA) in the synovial fluid and cartilage. Cell migration-inducing hyaluronidase 1 (CEMIP), also known as hyaluronan (HA)-binding protein involved in HA depolymerization (HYBID), was recently reported as an HA depolymerization-related molecule expressed in the cartilage of patients with OA. However, the underlying mechanism of CEMIP regulation is not well understood. We found that CEMIP expression was transiently increased by interleukine-1β (IL-1β) stimulation in chondrocytic cells. We also observed that ERK activation and NF-κB nuclear translocation were involved in the induction of CEMIP by IL-1β. In addition, both administration of HA and mechanical strain attenuated the CEMIP induction in IL-1β-stimulated chondrocytes. In conclusion, we clarified the regulatory mechanism of CEMIP in chondrocytes by inflammatory cytokines and suggested the potential involvement in osteoarthritis development.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-β-actina monoclonal antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-KIAA1199 antibody produced in rabbit, affinity isolated antibody