Merck
  • cDNA cloning and expression of a human platelet-derived growth factor (PDGF) receptor specific for B-chain-containing PDGF molecules.
cDNA cloning and expression of a human platelet-derived growth factor (PDGF) receptor specific for B-chain-containing PDGF molecules.
Molecular and cellular biology (1988-08-01)
L Claesson-Welsh, A Eriksson, A Morén, L Severinsson, B Ek, A Ostman, C Betsholtz, C H Heldin
RESUMEN
The structure of the human receptor for platelet-derived growth factor (PDGF) has been deduced through cDNA cloning. A 5.45-kilobase-pair cDNA clone predicts a 1,106-amino-acid polypeptide, including the cleavable signal sequence. The overall amino acid sequence similarity with the murine PDGF receptor is 85%. After transcription of the cDNA and translation in vitro, a PDGF receptor antiserum was used to immunoprecipitate a product of predicted size, which also could be phosphorylated in vitro. Stable introduction of the cDNA into Chinese hamster ovary (CHO) cells led to the expression of a 190-kilodalton component, which was immunoprecipitated by the PDGF receptor antiserum; this most probably represents the mature PDGF receptor. Binding assays with different 125I-labeled dimeric forms of PDGF A and B chains showed that the PDGF receptor expressed in CHO cells bound PDGF-BB and, to a lesser extent, PDGF-AB, but not PDGF-AA.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
PDGFRb active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)