Merck
The ubiquitin modifying enzyme A20 restricts B cell survival and prevents autoimmunity.
Immunity (2010-08-14)
Rita M Tavares, Emre E Turer, Chih L Liu, Rommel Advincula, Patrizia Scapini, Lesley Rhee, Julio Barrera, Clifford A Lowell, Paul J Utz, Barbara A Malynn, Averil Ma
RESUMEN
A20 is a ubiquitin modifying enzyme that restricts NF-kappaB signals and protects cells against tumor necrosis factor (TNF)-induced programmed cell death. Given recent data linking A20 (TNFAIP3) with human B cell lymphomas and systemic lupus erythematosus (SLE), we have generated mice bearing a floxed allele of Tnfaip3 to interrogate A20's roles in regulating B cell functions. A20-deficient B cells are hyperresponsive to multiple stimuli and display exaggerated NF-kappaB responses to CD40-induced signals. Mice expressing absent or hypomorphic amounts of A20 in B cells possess elevated numbers of germinal center B cells, autoantibodies, and glomerular immunoglobulin deposits. A20-deficient B cells are resistant to Fas-mediated cell death, probably due to increased expression of NF-kappaB-dependent antiapoptotic proteins such as Bcl-x. These findings show that A20 can restrict B cell survival, whereas A20 protects other cells from TNF-induced cell death. Our studies demonstrate how reduced A20 expression predisposes to autoimmunity.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-Actin Antibody, clone JLA20, clone JLA20, from mouse
Sigma-Aldrich
A20 human, recombinant, expressed in baculovirus infected Sf9 cells, ≥20% (SDS-PAGE)