Merck
  • Synthesis of Dinaphtho[2,3-d:2',3'-d']anthra[1,2-b:5,6-b']dithiophene (DNADT) Derivatives: Effect of Alkyl Chains on Transistor Properties.
Synthesis of Dinaphtho[2,3-d:2',3'-d']anthra[1,2-b:5,6-b']dithiophene (DNADT) Derivatives: Effect of Alkyl Chains on Transistor Properties.
International journal of molecular sciences (2020-04-05)
Takumi Ishida, Yuta Sawanaka, Ryota Toyama, Zhenfei Ji, Hiroki Mori, Yasushi Nishihara
RESUMEN
To investigate organic field-effect transistor (OFET) properties, a new thienoacene-type molecule, 4,14-dihexyldinaphtho[2,3-d:2',3'-d']anthra[1,2-b:5,6-b']dithiophene (C6-DNADT), consisting of π-conjugated nine aromatic rings and two hexyl chains along the longitudinal molecular axis has been successfully synthesized by sequential reactions, including Negishi coupling, epoxidation, and cycloaromatization. The fabricated OFET using thin films of C6-DNADT exhibited p-channel FET properties with field-effect mobilities (µ) of up to 2.6 × 10-2 cm2 V-1 s-1, which is ca. three times lower than that of the parent DNADT molecule (8.5 × 10-2 cm2 V-1 s-1). Although this result implies that the installation of relatively short alkyl chains into the DNADT core is not suitable for transistor application, the origins for the FET performance obtained in this work is fully discussed, based on theoretical calculations and solid-state structure of C6-DNADT by grazing incidence wide-angle X-ray scattering (GIWAXS) and atomic force microscopy (AFM) analyses. The results obtained in this study disclose the effect of alkyl chains introduced onto the molecule on transistor characteristics.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Etiniltrimetilsilano, 98%
Sigma-Aldrich
Pyridinium chlorochromate, 98%
Sigma-Aldrich
1,4-Benzenedimethanol, 99%
Sigma-Aldrich
Trimethylsulfonium iodide, 98%