Merck
Expanding the ChemGPS chemical space with natural products.
Journal of natural products (2005-07-26)
Josefin Larsson, Johan Gottfries, Lars Bohlin, Anders Backlund
RESUMEN
Recently various attempts have been made to increase the efficacy and precision of chemical libraries used in high-throughput screening (HTS) drug discovery approaches. One such approach is ChemGPS, which provides a defined chemical space for prescreening evaluation of chemical compound properties or virtual dereplication. In the present study, ChemGPS has been applied to a set of natural products shown to exhibit cyclooxygenase-1 and/or -2 (COX-1/2) inhibition. With the purpose of defining chemical properties and linking these to the observed mode of enzyme inhibition, this resulted in two lines of reasoning. On one hand several specific features of these compounds have been identified and discussed. Overall COX inhibition was frequently correlated with the presence of at least one ring in the structure, fragments exhibiting structural rigidity, and a relatively large molecular size. The concept "size" includes several parameters, e.g., molecular volume, weight, and number of bonds. On the other hand, and possibly even more important, was the unexpected finding that the natural products studied to a large extent fell outside the defined ChemGPS chemical space. Therefore, we also propose an expanded space for natural products: ChemGPS-NP.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Sigma-Aldrich
Zimtaldehyd, natural, ≥95%, FG
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Ácido linoleico, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
trans-Cinnamaldehyde, 97%
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
L-Menthol, natural, ≥99%, FCC, FG
Sigma-Aldrich
Oleanolic acid, ≥97%
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Sigma-Aldrich
Rosmarinic acid, 96%
Sigma-Aldrich
Capsaicin, natural
Sigma-Aldrich
Quercetin 3-β-D-glucoside, ≥90% (HPLC)
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
trans-Cinnamaldehyde, ≥99%
Sigma-Aldrich
L-Menthol, ≥99%, FCC, FG