Merck
Aberrant Differentiation of Human Pluripotent Stem Cell-Derived Kidney Precursor Cells inside Mouse Vascularized Bioreactors.
Nephron (2020-08-07)
Parisa Ranjzad, Jessica Jinks, Amir P Salahi, Ioannis Bantounas, Brian Derby, Susan J Kimber, Adrian S Woolf, Jason K F Wong
RESUMEN
Numerous studies have documented the in vitro differentiation of human pluripotent stem cells (hPSCs) into kidney cells. Fewer studies have followed the fates of such kidney precursor cells (KPCs) inside animals, a more life-like setting. Here, we tested the hypothesis that implanting hPSC-derived KPCs into an in vivo milieu surgically engineered to be highly vascular would enhance their maturation into kidney tissues. 3D printed chambers containing KPCs were implanted into the thighs of adult immunodeficient mice. In some chambers, an arterial and venous flow-through (AVFT) was surgically fashioned. After 3 weeks and 3 months, implants were studied by histology, using qualitative and quantitative methods. After 3 weeks, chambers containing AVFTs were richer in small vessels than contralateral chambers without AVFTs. Glomeruli with capillary loops and diverse types of tubules were detected in all chambers. At 3 months, chambers contained only rudimentary tubules and glomeruli that appeared avascular. In chambers with AVFTs, prominent areas of muscle-like cells were also detected near tubules and the abnormal tissues immunostained for transforming growth factor β1. These features have similarities to renal dysplasia, a typical histological signature of human congenital kidney malformations. This study urges a note of caution regarding the in vivo fates of hPSC-derived kidney precursors, with pathological differentiation appearing to follow a period of increased vascularity.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-actina, α-músculo liso monoclonal, clone 1A4, ascites fluid
Sigma-Aldrich
Anticuerpo anti-mitocondrial, superficie de mitocondrias intactas, clon 113-1, clone 113-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Podocin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution